Learn More
Medchemexpress LLC H2-D1 Mouse His-SU 20ug

Supplier: Medchemexpress LLC HYP7151820UG
Recombinant mouse H2-D1 protein (residues G25-T311) expressed in E. coli and provided lyophilized with an N-terminal His-SUMO tag. Intended for immunology and antigen presentation research, the material is supplied with a datasheet and certificate of analysis, and formulated for stability during storage.
- Recombinant sequence matching residues G25-T311 for relevant biological studies.
- Expressed in E. coli for scalable production and consistent quality.
- N-terminal His-SUMO tag enables affinity purification and improved solubility.
- Purity greater than 90% by reducing SDS-PAGE with supporting certificate of analysis.
- Lyophilized formulation with trehalose for enhanced stability during storage.
- Available in microgram-scale vials suitable for biochemical and immunological assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.