Learn More
Sino Biological Influenza A H3N2 (A/Massachusetts/18/2022; A/Croatia/10136RV/2023) Neuraminidase / NA Protein (ECD, His Tag)
Supplier:  Sino Biological 40991V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Massachusetts/18/2022; A/Croatia/10136RV/2023 (H3N2)) neuraminidase (translated amino acids of EPI2096147) (Ile30-Thr469) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3346