Learn More
Sino Biological H3N2 NP PAb 100ul (Sino Biological)
Supplier:  Sino Biological 40208T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein / NP ( Catalog#40208-V08B; P22435.2; Met1-Asn498). Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein / NP specific IgG was purified by Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein / NP affinity chromatography.
Catalog No. 50-237-3744