Learn More
ATCC H4; Neuroglioma; Human (Homo sapiens)
Supplier:  ATCC HTB148
Encompass
H4 is an epithelial cell line that was isolated from the brain of a 37-year-old, White male with neuroglioma. The cell line H4 can be used in neuroscience research.
Catalog No. 50-238-3279