Learn More
ATCC H9c2(2-1); Embryonic Myocardium; Rat (Rattus norvegicus)
Supplier:  ATCC CRL1446
Encompass
H9c2(2-1) is a subclone of the original clonal cell line derived from embryonic BD1X rat heart tissue that exhibits many of the properties of skeletal muscle. This cell line can be used in cardiovascular disease research.
Catalog No. 50-238-2220