Learn More
Abcam HA 100 dihydrochloride, Myosin light chain kinase, PKA, PKC and PKG inhibitor, 10MG
Supplier:  Abcam AB14556710MG
Encompass_Preferred
MW 313.8 Da, Purity >98%. HA 100 dihydrochloride, Myosin light chain kinase, PKA, PKC and PKG inhibitor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5138