Learn More
Enzo Life Sciences HA.11 polyclonal antibody (purified) (0.5ml)
SDP
Supplier:  Enzo Life Sciences ENZABS2030500
Encompass_Preferred
Immunogen: Twelve amino acid peptide CYPYDVPDYASL. Specificity: Recognizes the influenza hemagglutinin epitope (YPYDVPDYA). Applications: IP, WB. Formulation: Liquid. In PBS containing 0.03% thimerosal.
Catalog No. 50-201-2753