Learn More
CPC Scientific H-Cys-Tyr-Pro-Tyr-Asp-Val-Pro-Asp-Tyr-Ala-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific TAGP003A
Encompass
This 10-amino acid peptide epitope tag from hemophilus influenza is recognized by the common monoclonal antibody 12Ca5.ONE-LETTER SEQUENCE: CYPYDVPDYAMOLECULAR FORMULA: C56H72N10O18S1MOLECULAR WEIGHT:1205.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: ImmunomodulationREFERENCES: Sumi, T. et al. J. Cell Biol. 147, 1519 (1999); Xie, L. et al. Endocrinol. 139, 4563 (1998).
Catalog No. 50-211-0532