Learn More
TA Instruments HA Aluminum Helical Cup, Diameter 34 mm, Height 50 mm, for Use with Stainless Steel Helical Rotor HR X0, DHR and AR-G2 P/N 546018.901 or AR P/N 546019.901, and Smart Swap Concentric Cylinder Peltier Jacket P/N 533201.901
Supplier:  TA Instruments 546093.001
Encompass_Preferred
Waters TA Instruments HA Aluminum Helical Cup has a diameter of 34 mm and a height of 50 mm. It is used with HR X0, DHR, and AR-Series Smart Swap Concentric Cylinder Peltier Jacket (P/N 533201.901) and either the Stainless Steel Helical Rotor for HR X0, DHR and AR-G2 (P/N 546018.901) or AR (P/N 546019.901).
Catalog No. 50-005-80764