Learn More
Santa Cruz Biotechnology HA PROBE F-7 MOUSE MONOCLONAL
Supplier:  Santa Cruz Biotechnology SC7392
Encompass
HA-probe (F-7), mouse monoclonal IgG2a; 200microgram/ml, epitope mapping to an internal region of the influenza hemagglutinin (HA) protein, specific for proteins containing the HA tag, Western blotting, immunoprecipitation and immunohistoch
Catalog No. NC9682981