Learn More
Santa Cruz Biotechnology HA-PROBE (F7AC)
Supplier:  Santa Cruz Biotechnology SC7392 AC
Encompass
HA-probe (F-7) agarose conjugate, mouse monoclonal IgG2a; 200microgram/ml, epitope mapping to an internal region of the influenza hemagglutinin (HA) protein, specific for proteins containing the HA tag, Western blotting
Catalog No. NC9778567