Learn More
Abcam HA14-1, Bcl-2 inhibitor. BH-3 mimetic., 10MG
Supplier:  Abcam AB14130610MG
Encompass_Preferred
MW 409.2 Da. Cell permeable Bcl-2 inhibitor. BH-3 mimetic. Disrupts Bax and Bcl-2 interactions. Apoptotic anticancer agent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6698