Learn More
Hach Company Hach Hypochlorite NH3 Reagent, 500 mL (52007)
Supplier:  Hach Company 52007
Encompass
Hypochlorite NH3 reagent is a sodium hypochlorite-based prepared reagent supplied in a 500 mL bottle for laboratory ammonia analysis by flow injection analysis (FIA). It is intended for routine laboratory testing; follow method recommendations for opened reagent use.
  • Sodium hypochlorite reagent suitable for ammonia analysis using FIA.
  • Volume: 500 mL.
  • Prepared reagent intended for laboratory use and compatible with ammonia reagent sets.
  • Shelf life: 3 months unopened; consult method recommendations after opening.
  • Store and handle according to safety data and laboratory procedures for hypochlorite solutions.
Catalog No. NC3804635