Learn More
Medchemexpress LLC HY-131045 5mg Medchemexpress, HADA (hydrochloride) CAS:2253733-10-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1310455MG
Encompass_Preferred
Medchemexpress, HY-131045 5mg HADA (hydrochloride) CAS:2253733-10-5 HADA hydrochloride (HCC-Amino-D-alanine hydrochloride) is a blue (λem~450 nm) fluorescent D-amino acid (FDAA). FDAAs are efficiently incorporated into the peptidoglycans (PGs) of diverse bacterial species at the sites of PG biosynthesis, allowing specific and covalent probing of bacterial growth with minimal perturbation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1260