Learn More
Strem, An Ascensus Company CAS# 17475-67-1. 5g. Hafnium(IV) acetylacetonate, min. 96%. MFCD00044997
SDP
Supplier:  Strem, An Ascensus Company 7220025G
Encompass
CAS# 17475-67-1. 5g. Hafnium(IV) acetylacetonate, min. 96%. MFCD00044997. Molecular Weight: 574.93. Molecular Formula: Hf(CH3COCHCOCH3)4. Color/form: white to off-white pwdr. Strem# 72-2002. http://www.strem.com/catalog/v/72-2002/
Catalog No. 50-901-12945