Learn More
Analytical Sales & Services Half-Height Reservoir 12 Channel, 170 mL Total Volume, 12-Channel Design, Half Height Profile
Supplier:  Analytical Sales & Services 47170
Encompass
The Half-Height Reservoir, 12 Channel, manufactured by Analytical Sales & Services, enhances laboratory efficiency with its optimized design for liquid handling. Featuring a pyramid bottom and 12 channels, it minimizes reagent waste while maximizing throughput for multi-channel pipetting. Constructed from clear polypropylene for chemical compatibility, it allows for unobstructed visibility of liquid levels, making it ideal for automated workflows. Total volume is 170 mL, reducing the need for frequent refills.
Catalog No. NC0719606