Learn More
Advanced Materials Technology HALO 160 A C30 2.7 um 4.6 x 150 mm
Supplier:  Advanced Materials Technology 92114730
Encompass_Preferred
HALOC30 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in 2.7 µm particle size and a wide range of column dimensions, HALO 160 Å C30 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. NC3596261