Learn More
Advanced Materials Technology HALO 160 C30 2.7 2.1 X
Supplier:  Advanced Materials Technology 92112130
Encompass_Preferred
HALO C30 columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in 2.7 µm particle size and a wide range of column dimensions, HALO 160 Å C30 columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-255-6566