Learn More
Advanced Materials Technology HALO 160 PHENYL-HEXYL 2.7
Supplier:  Advanced Materials Technology 91215706
Encompass_Preferred
HALO BIOCLASS Phenyl-Hexyl Peptide columns made with innovative Fused-Core particle technology are ideal for both ultrafast and ultrahigh resolution separations of peptides and polypeptides up to 20 kDa. HALO peptide columns offer comparable peak capacity to sub-2 μm non-core columns at 40–50% back pressure (2.7 μm). HALO 160 Å peptide columns are optimized for high performance in HPLC, UHPLC, and LCMS peptide applications.
Catalog No. 50-255-6734