Learn More
Advanced Materials Technology HALO 90 GLYCAN 2.7 2.1
Supplier:  Advanced Materials Technology 92922405
Encompass_Preferred
HALO Glycan specific solution is ideal for HILIC separations. Offers high resolution even with complex glycan mixtures
Catalog No. 50-255-6096