Learn More
Advanced Materials Technology HALO 90 PAH 2.7 4.6 X 5
Supplier:  Advanced Materials Technology 92844412
Encompass_Preferred
The HALO PAH column is an un-endcapped trifunctional C18 bonded phase with a proprietary manufacturing process designed on proven Fused-Core technology to provide a fast, efficient separation of PAH compounds. Best Applications: EPA 610, EPA 8310, EU 15+1
Catalog No. 50-255-6102