Learn More
Advanced Materials Technology HALO 90 Å HILIC, 2.7 µm, 2.1 x 50 mm
Supplier:  Advanced Materials Technology 92812401
Encompass_Preferred
HALO HILIC columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å HILIC columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-900-02090