Learn More
Advanced Materials Technology HALO 90A PCS C18, 2.7um 4.6 mm x 50 mm
Supplier:  Advanced Materials Technology 92814417
Encompass_Preferred
Built upon proven Fused-Core technology for speed and efficiency, the HALO PCS column products are positively charged surface chemistries designed to deliver improved peak shapes for basic compounds observed with standard C18 chemistry. Ideal for use with low ionic strength mobile phases, HALO PCS maintains peak symmetry at higher loading capacities and provides alternate selectivities from C18. Available in both a 90 Å and 160 Å pore size for small molecule and peptide analysis. The columns are optimized to deliver performance for reproducible, high efficiency LC and LCMS separations.
Catalog No. NC2505053