Learn More
Advanced Materials Technology HALO 90 Å PFP, 2.7 µm, 3.0 x 100 mm
Supplier:  Advanced Materials Technology 92813609
Encompass_Preferred
HALO PFP columns made with innovative Fused-Core particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å PFP columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
Catalog No. 50-752-8157