Learn More
Medchemexpress LLC Halofenozide | 112226-61-6 | 99.4% | 330.81 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11389010MM/1ML
Encompass_Preferred
Halofenozide (RH-0345) is an ecdysteroid agonist belonging to a new group of insect growth regulators (IGRs) that mimic the action of the natural insect molting hormone 20-hydroxyecdysone. It is intended for research use only.
  • Reduces growth and development of oocytes in vitro.
  • Significantly reduces oviposition period, fecundity, and egg viability in adult females (in vivo, at higher doses).
  • Causes smaller eggs with decreased length, width, and volume (in vivo).
Catalog No. 50-004-37804