Learn More
Cayman Chemical HalofugInonhydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 133705 MG
Encompass_Preferred
A halogenated derivative of febrifugine that down-regulates Smad3, blocking TGF-β signaling relevant to fibrosis and tumor progression in a variety of different models; competitively inhibits prolyl-tRNA synthetase (Ki = 18.3 nM), preventing the differentiation of TH17 cells and blunting an autoimmune response
Catalog No. 50-295-5260