Learn More
Cayman Chemical HaloperIdol 500mg
SDP
Supplier:  Cayman Chemical 12014500 MG
Encompass_Preferred
A typical antipsychotic and dopamine D2-like receptor antagonist (Kis = 0.6, 0.2, and 22 nM, for D2, D3, and D4 receptors, respectively); An inverse agonist at D2 and D3 receptors (IC50s = 0.8 and 0.6 nM, respectively); binds to α1- and α2- adrenergic and histamine H1 receptors, as well as 5-HT receptor subtypes 5-HT1D and 5-HT2A (Kds = 17, 600, 260, 40, and 61 nM, respectively); inhibits stereotypic behavior induced by apomorphine and amphetamine in rats (ID50s = 0.532 and 0.101 μmol/kg, respectively); inhibits apomorphine-induced decreases in prepulse inhibition of the acoustic startle response in rats in a dose-dependent manner
Catalog No. 50-295-2296