Learn More
Apexbio Technology LLC Haloperidol 52-86-8 500mg
SDP
Supplier:  Apexbio Technology LLC B2099500
Encompass_Preferred
Haloperidol is a small-molecule antagonist targeting dopamine receptors It is designed to block dopamine signaling thereby modulating neurotransmission within cortical limbic and nigrostriatal pathways Haloperidol exerts its biological activity primarily through inverse agonism at dopamine receptors Based on these pharmacological properties Haloperidol holds research potential in psychiatric and neuropsychological applications including studies of delirium delusions hallucinations and characterization of dopamine-mediated neuronal circuits and behavioral models in psychiatric disorders
Catalog No. 50-000-92311