Learn More
Cayman Chemical HaloperIdol-d4 1mg
SDP
Supplier:  Cayman Chemical 261161 MG
Encompass_Preferred
An internal standard for the quantification of haloperidol by GC- or LC-MS
Catalog No. 50-293-2741