Learn More
Medchemexpress LLC Haloperidol decanoate | 74050-97-8 | MFCD00866706 | 99.7% | 530.11 | C31H41ClFNO3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10796925MG
Encompass_Preferred
Haloperidol decanoate is the decanoate ester of haloperidol, provided as a research reagent for studies of antipsychotic pharmacology and dopamine receptor activity. It is formulated as a long-acting depot ester commonly used in in vivo and analytical investigations of sustained antipsychotic effects.
  • Long-acting depot ester for antipsychotic pharmacology studies.
  • High purity suitable for analytical and in vivo research (reported ~99.7%).
  • Molecular formula C31H41ClFNO3 and molecular weight 530.11 for accurate dosing and calculation.
  • Available in small mg pack sizes convenient for research workflows.
Catalog No. 50-002-41946