Learn More
Medchemexpress LLC Haloperidol (Standard) | 52-86-8 | 99.8% | C₂₁H₂₃ClFNO₂ | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14538R100MG
Encompass_Preferred
Haloperidol (Standard) is an analytical standard of Haloperidol. It functions as a potent dopamine D2 receptor antagonist, commonly used as an antipsychotic. This product is intended for various research and analytical applications.
  • Potent dopamine D2 receptor antagonist
  • Used as an antipsychotic
  • Analytical standard for research
  • Utilized in qualitative, quantitative, and methodological experiments
  • Applicable for HPLC, GC, and MS
Catalog No. 50-003-55808