Learn More
Medchemexpress LLC Haloperidol (standard) | 52-86-8 | MFCD00051423 | 99.84% | C₂₁H₂₃ClFNO₂ | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14538R50MG
Encompass_Preferred
Haloperidol (Standard) is the analytical standard of Haloperidol, intended for research and analytical applications. It is a potent dopamine D2 receptor antagonist, widely used as an antipsychotic. This compound is of analytical standard grade, used as a reference standard for assay.
  • Used in qualitative, quantitative and methodological research experiments
  • Suitable for HPLC, GC and MS applications
Catalog No. 50-003-53950