Learn More
Bioworld Ham's F-10 Nutrient Mix w/ L-Glut, 1 L
Supplier:  Bioworld 306110361
Encompass_Preferred
Ham's Nutrient Mixtures, F-10 and F-12, were both originally developed to support the growth of a variety of cells, particularly the Chinese Hamster Ovary (CHO) cells, with or without serum supplementation. Ham's F-10 has since been used in the culture of a wide variety of cell of human and animal origin. Ham's F-12 is preferred by many for the culture of rodent cells. It has also shown excellent results as a cloning medium for myeloma and hybridoma cells. Ham's F-10 Nutrient Mixtures, item # 785081, powder includes L-glutamine. Does not include sodium bicarbonate. Click here to see a Sodium Bicarbonate addition table for powdered media. Click here to see a Ham's F-10 Nutrient Components Table.
Catalog No. 50-254-092