Learn More
Alpco Ltd Human Anti Mouse Antibodies ELISA (HAMA ELISA)
SDP
Supplier:  Alpco Ltd 43HAMHUE01
Encompass_Preferred
The Human Anti Mouse Antibodies ELISA (HAMA ELISA) is produced for the quantitative determination of human anti-mouse IgG antibody (HAMA) levels in serum or plasma samples. It detects both HAMA-IgG and HAMA-IgM subtypes. The Human Anti Mouse Antibodies ELISA (HAMA ELISA) might be used as an aid for detection of subjects with positive HAMA that may affect research involving monoclonal murine IgG. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-922-300