Learn More
Sigma Aldrich Fine Chemicals Biosciences Hamilton PRP 1 HPLC Column
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 82016U
Encompass
The Hamilton PRP-1 polymeric reversed phase columns are Ideal for high pH (pH 8-13) operation or with analytes that give poor chromatogrpahy on silica-based columns.
Catalog No. 50-290-8998