Learn More
Hamilton™ Threaded Plunger Syringes
Supplier:  Hamilton™ 81141
Catalog No. 14-815-328
Description
Description
For minute fluid manipulations; Luer tip syringe terminations.
For applications requiring extremely precise plunger movement or minute fluid manipulation. Dispense from 0.33 to 151.14μL per plunger revolution. Luer tip (LT) syringe terminations for easy connection of Kel-F™ hub, stainless-steel needles; Kel-F hub PTFE-resin tubing; and 1/4 O.D. x 1/8 in. I.D. Tygon™ or rubber tubing.
Product Suggestions
Product Suggestions