Learn More
MyBioSource Hamster Triglyceride Lipase (TGL) ELISA Kit
SDP
Supplier:  MyBioSource MBS025126
Encompass_Preferred
The Hamster TGL ELISA Kit quantitatively measures TGL in various biological samples using a double-antibody sandwich assay. It has a 0.156-10 ng/ml detection range and includes reagents for 96 wells, with a total assay time of about 3.5 hours.
Catalog No. NC2831172