Learn More
Grainger Hand Crank Hose Reel 100 ft 3/8 ID Blue
Supplier:  Grainger 16X555
Encompass_Preferred
The Coxreels Hand Crank Hose Reel is a 100 ft capacity steel hose reel with powder coat finish. It features a slip ring bracket to prevent hose wear and has a hand crank rewind with fold down crank handle and adjustable hose guide. This industrial grade hose reel can hold 3/4" ID hose and has a max pressure rating of 300 PSI. It comes with mounting hardware for wall or ceiling installation. The reel dimensions are 23"W x 23"H x 12"D and it weighs 35 lbs.
Catalog No. 50-276-7143