Learn More
Laboratory Supply Network Inc HANDHELD HOMOGENIZER
SDP
Supplier:  Laboratory Supply Network Inc D1000
Encompass_Preferred
Benchmark's new handheld homogenizer, the D1000, is designed to rapidly homogenize, emulsify, suspend and disrupt biological samples. Ideal for use with microtubes, the homogenizer works well with samples from 0.1ml to 250ml (depending upon the generator probe used).
Catalog No. NC1299259