Learn More
Avantor Fluid Handling Handheld Remote Controller; DB25(M) Connector; 6 ft (1,8 m) Cable
Supplier:  Avantor Fluid Handling MFLX0752397
Encompass_Preferred
Easily control your Masterflex® L/S® pumps while filling and dispensing.Suitable for use with Masterflex® L/S® pumpsThese ergonomic handheld controllers are designed to control basic pump drive operations including start/stop, direction, and prime — depending on available drive functions. Route tubing through the handle for convenience in filling and dispensing applications. These handheld controllers accept Masterflex tubing sizes L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18, L/S 15, and L/S 24 (up to 7/16" nominal OD).
Catalog No. NC3316498