Learn More
Hoefer, Inc HandheldCap 8 Capper/Decapper
SDP
Supplier:  Hoefer, Inc STHCM8
Encompass_Preferred
User-friendly and lightweight, Hoefer HandheldCap 8/6 can quickly cap/decap 8-ch or 6-ch Hoefer sample storage tubes, effectively reducing the work load of operators as a solution that takes into account the transition from manual to automated systems.Features and Benefits:Automatic capping and decapping within 10 seconds, improving the processing throughput by handheld. User-friendly, lightweight and ergonomically designed. Magnetic contact charging for longer battery life and faster charging times. Portable and convenient, a better choice for sample treatment in any laboratory. Desktop stand for easy handling, firmly attached.
Catalog No. NC2956935