Learn More
Sigma Aldrich Fine Chemicals Biosciences Hanks Balanced Salt solut500ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H66486X500ML
Encompass
The essential function of Hanks Balanced Salt SOLUTIONon is to maintain pH as well as osmotic balance. It also provides water and essential inorganic ions to cells.
Catalog No. 50-290-0167