Learn More
ATCC Hank's BSS w/o Ca, Mg, or P Red; 500 ML
Supplier:  ATCC 302213
Encompass
Hank's Balanced Salt Solution (HBSS) is used for washing cells, for use as a diluent, or for use as an inorganic base for standard media preparation. It has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-4257
 Restricted Item