Learn More
Medchemexpress LLC HAP-1 | 329004-38-8 | 99.1% | 1335.47 | 100 MG

Supplier: Medchemexpress LLC HYP4096100MG
HAP-1 is a synovial-targeted transduction peptide that facilitates specific internalization of protein complexes into human and rabbit synovial cells. It has been fused to an antimicrobial peptide, (KLAK)2, to create a proapoptotic peptide called DP2.
- Facilitates specific internalization of protein complexes into human and rabbit synovial cells.
- Can be fused to an antimicrobial peptide, (KLAK)2, to create a proapoptotic peptide DP2.
- Shipping at room temperature in continental US; may vary elsewhere.
- Store sealed, away from moisture.
- Powder can be stored at -80°C for 2 years or -20°C for 1 year.
- In solvent, store at -80°C for 6 months or -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.