Learn More
Medchemexpress LLC HAP-1 | 329004-38-8 | 99.1% | 1335.47 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP4096
Encompass_Preferred
HAP-1 is a synovial-targeted transduction peptide designed to facilitate the specific internalization of protein complexes into human and rabbit synovial cells. It can be fused to antimicrobial peptides, such as (KLAK)2, to generate proapoptotic peptides. This product is for research use only.
  • Synovial-targeted transduction peptide
  • Facilitates specific internalization of protein complexes into human and rabbit synovial cells
  • Can be fused to antimicrobial peptides to generate proapoptotic peptides
Catalog No. 50-003-98969