Learn More
Essendant HARDHAT10/CTWH
Supplier:  Essendant MMMX5001VANSI
Encompass_Preferred
3M SecureFit X5000 Series Safety Helmets provide workers with a comfortable secure fit that s ideal for a variety of applications Climbing helmet-inspired brimless design with chin strap meets occupational safety requirements while helping the wearer maintain greater situational awareness by delivering an obstruction-free field of vision Low-sitting suspension system minimizes pressure to the head while its exclusive patented Pressure Diffusion Technology allows the system to self-adjust to the wearer as the smooth easy-turning ratchet is tightened Multiple levels of height and head-shape adjustments allow the wearer to customize the fit even further X5000 Series helmets meet ANSI safety compliance standards and are designed to protect the wearer s head from small falling objects All helmets are compatible with a variety of 3M face-shields earmuffs and additional accessories (all sold separately) Vented Air-circulation vents along the helmet s crown help keep workers coo
Catalog No. 50-000-35916