Learn More
Hach Company Hardness 3 Solution, 500 mL
Supplier:  Hach Company 42649
Encompass
EDTA titrant solution for hardness determination by drop-count titration. 0.03 N (0.015 M) 500 mL bottle
Catalog No. 50-199-4288