Learn More
Cayman Chemical HArgGlyAspCysOHtrIfluo 10mg
SDP
Supplier:  Cayman Chemical 3454310 MG
Encompass_Preferred
An RGD-containing tetrapeptide; inhibits the binding of fibrinogen to endothelial cells and ADP-stimulated platelets (IC50s = 320 and 35 µM, respectively); implantation of titanium rods coated with H-Arg-Gly-Asp-Cys-OH increases bone formation in rat femurs; has been conjugated to polyethylenimine to improve gene transfection efficiency
Catalog No. 50-295-0033