Learn More
Medchemexpress LLC Harmine (Standard) | 442-51-3 | 212.25 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0737AR
Encompass_Preferred
Harmine (Standard) is the analytical standard of Harmine, intended for research and analytical applications. It is a natural dual-specificity tyrosine phosphorylation-regulated kinase (DYRK) inhibitor with anticancer and anti-inflammatory activities, exhibiting high affinity for the 5-HT2A serotonin receptor with a Ki of 397 nM. The compound is a reference standard supplied assay, commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications.
  • Natural dual-specificity tyrosine phosphorylation-regulated kinase (DYRK) inhibitor.
  • Possesses anticancer and anti-inflammatory activities.
  • High affinity for 5-HT2A serotonin receptor (Ki of 397 nM).
  • Reference standard supplied assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
Catalog No. 50-003-90223