Learn More
Medchemexpress LLC Harmine (Standard) | 442-51-3 | 212.25 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYN0737AR
Encompass_Preferred
Harmine (Standard) is the analytical standard of Harmine, intended for research and analytical applications. It is a natural dual-specificity tyrosine phosphorylation-regulated kinase (DYRK) inhibitor with anticancer and anti-inflammatory activities. Harmine also exhibits a high affinity for the 5-HT2A serotonin receptor, with a Ki of 397 nM.
  • Analytical standard for Harmine.
  • Intended for research and analytical applications.
  • Natural dual-specificity tyrosine phosphorylation-regulated kinase (DYRK) inhibitor.
  • Possesses anticancer and anti-inflammatory activities.
  • High affinity for 5-HT2A serotonin receptor (Ki of 397 nM).
  • Used in HPLC, GC, and MS for qualitative, quantitative, and methodological research.
Catalog No. 50-003-89938